
4-sec-Butylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAB4004-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:155.29 g/mol
Boiling Pt:105°C
UN:2735
ADR:8,III
:GHS05
CAS Number:25834-93-9
Formula:C₁₀H₂₁N