N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:74844-91-0
:
Formula:C₁₁H₁₉NO₅
MDL Number:MFCD00076981
Boiling Pt:132°C
:245.28 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:94.0 to 98.0 deg-C
- Specific rotation [a]20/D:-77.0 to -80.0 deg (C=1, methanol)
Specification Test Results
Related Information
Frequently Bought Together