N-(tert-Butoxycarbonyl)-N-methyl-D-alanine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19914-38-6
:
Formula:C₉H₁₇NO₄
MDL Number:MFCD00063137
:203.24 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:90.0 to 94.0 deg-C
- Specific rotation [a]20/D:+29.0 to +31.0 deg (C=1, ethanol)