(1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥97.0% (by GC, titration analysis)
Catalog # TCIAB3538-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 97.0 %
- Specific rotation [a]20/D:-48.0 to -52.0 deg (C=1, MeOH)
Specification Test Results
About this item
:
:243.30 g/mol
CAS Number:222530-39-4
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD04973107