2-(3-Bromopropyl)-1,3-dioxolane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:62563-07-9
:
Formula:C₆H₁₁BrO₂
MDL Number:MFCD09868694
:195.06 g/mol
Boiling Pt:88°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together