Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-3-methyl-2-butanone
3-Bromo-3-methyl-2-butanone ≥96.0% (by GC)
3-Bromo-3-methyl-2-butanone ≥96.0% (by GC)
Flammable
Irritant
3-Bromo-3-methyl-2-butanone ≥96.0% (by GC)
Supplier:
TCI
Certificates
Expand image
Flammable
Irritant
3-Bromo-3-methyl-2-butanone ≥96.0% (by GC)
Supplier:
TCI
About this item
Specification Test Results
Appearance:
Colorless to Yellow clear liquid
Purity (GC):
min. 96.0 %
Specification Test Results