4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:130309-46-5
:
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD03453262
:243.30 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 % (total of isomer)
- Purity (Neutralization titration):min. 98.0 %