Order Entry
Finland
ContactUsLinkComponent
1,3-Bis[(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)
1,3-Bis[(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)
Catalog # TCIAB3216-1G
Supplier:  TCI
CAS Number:  
1,3-Bis[(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)
Catalog # TCIAB3216-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    58.0 to 62.0 deg-C

Specification Test Results

About this item

 :
 :270.52 g/mol
Storage Temperature:Refrigerator
CAS Number:38170-80-8
Formula:C₁₆H₂₂Si₂
MDL Number:MFCD00078305