Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biotin-PEG3-amine
Biotin-PEG3-Amine ≥95.0% (by HPLC)
Biotin-PEG3-Amine ≥95.0% (by HPLC)
:
TCI
Biotin-PEG3-Amine ≥95.0% (by HPLC)
:
TCI
:
359860-27-8
:
:
C₁₈H₃₄N₄O₅S
:
MFCD09952640
:
Freezer
:
418.55 g/mol
Appearance:
White to Almost white powder to crystal
Purity (HPLC):
min. 95.0 area%
Elemental analysis (Nitrogen):
12.30 to 14.30 %