Order Entry
Finland
ContactUsLinkComponent
4,4'-Biphenyldiboronic acid bis(neopentyl glycol)ester ≥98.0% (by GC, titration analysis)
4,4'-Biphenyldiboronic acid bis(neopentyl glycol)ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3151-1G
Supplier:  TCI
CAS Number:  
4,4'-Biphenyldiboronic acid bis(neopentyl glycol)ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3151-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :378.0773 g/mol
CAS Number:5487-93-4
Formula:C₂₂H₂₈B₂O₄
MDL Number:MFCD01318103