Order Entry
Finland
ContactUsLinkComponent
 
N-(tert-Butoxycarbonyl)-D-threonine ≥98.0% (by HPLC, titration analysis)
  TCIAB2990-5G
 :  TCI
 :  
N-(tert-Butoxycarbonyl)-D-threonine ≥98.0% (by HPLC, titration analysis)
  TCIAB2990-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +8.0 to +11.0 deg (C=1, AcOH)

 

 

 :
 :219.24 g/mol
 :55674-67-4
 :C₉H₁₇NO₅
 :MFCD00037807