
1,10-Bis(acryloyloxy)decane ≥90.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:13048-34-5
:
Formula:C₁₆H₂₆O₄
MDL Number:MFCD00080453
:282.38 g/mol
Boiling Pt:181°C
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:235-922-4
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 90.0 %