Order Entry
Finland
ContactUsLinkComponent
trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one ≥98.0% (by HPLC)
trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
trans,trans-1,5-Bis(4-fluorophenyl)-1,4-pentadien-3-one ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    152.0 to 156.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together