Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(+/-)-alpha-Bisabolol
(+/-)-ɑ-Bisabolol ≥80.0% (by GC)
(+/-)-ɑ-Bisabolol ≥80.0% (by GC)
(+/-)-ɑ-Bisabolol ≥80.0% (by GC)
TCIAB2119-25ML
:
TCI
:
(+/-)-ɑ-Bisabolol ≥80.0% (by GC)
TCIAB2119-25ML
:
TCI
:
Pk:
25 mL
Seq:
2315
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 80.0 %
:
:
222.37 g/mol
:
14,01241
:
208-205-9
:
515-69-5
:
C₁₅H₂₆O
:
MFCD00129082