

2-[4-(Bromomethyl)phenyl]propionic acid ≥98.0% (by titrimetric analysis)
: TCI
:111128-12-2
:
:C₁₀H₁₁BrO₂
:MFCD02093445
:243.10 g/mol
:GHS07
:GHS08
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:125.0 to 131.0 deg-C
Frequently Bought Together