
9-(Bromomethyl)acridine ≥98.0% (by HPLC, titration analysis) for HPLC labeling
Supplier: TCI
Certificates
About this item
CAS Number:1556-34-9
:
Formula:C₁₄H₁₀BrN
MDL Number:MFCD01321146
:272.15 g/mol
Melting Pt:165°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %