
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) ≥98.0% (by GC) triflating reagent
Catalog # TCIAB1871-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:38.0 to 42.0 deg-C
Specification Test Results
About this item
:
:358.23 g/mol
:GHS07
CAS Number:145100-50-1
Formula:C₇H₄F₆N₂O₄S₂
MDL Number:MFCD00191834