Order Entry
Finland
ContactUsLinkComponent
N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone ≥98.0% (by HPLC, titration analysis)
N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information