Bis(2-formylphenyl)ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:49590-51-4
:
Formula:C₁₄H₁₀O₃
MDL Number:MFCD00191616
Boiling Pt:182°C
:226.23 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:77.0 to 80.0 deg-C
- Solubility in Methanol:almost transparency