Boc-trans-4-hydroxy-L-proline ≥98.0% (by titrimetric analysis)
Catalog # TCIAB1635-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-67.0 to -69.0 deg (C=1, MeOH)
- Solubility in Water:very faint turbidity
Specification Test Results
About this item
:
:231.3 g/mol
CAS Number:13726-69-7
Formula:C₁₀H₁₇NO₅
MDL Number:MFCD00053370