
Phenacyl-4-(bromomethyl)phenylacetate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:66270-97-1
:
Formula:C₁₇H₁₅BrO₃
MDL Number:MFCD00009632
:347.21 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:87.0 to 89.0 deg-C
- Solubility in Toluene:very faint turbidity