
4,5-Bis(mercaptomethyl)-o-xylene ≥96.0% (by GC, titration analysis)
Catalog # TCIAB1105-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Iodometric Titration):min. 96.0 %
- Melting point:65.0 to 69.0 deg-C
Specification Test Results
About this item
:
:198.34 g/mol
EINECS:233-551-2
:GHS07
CAS Number:10230-61-2
Formula:C₁₀H₁₄S₂
MDL Number:MFCD00004869