Order Entry
Finland
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)acetamide kit ≥80.0% (by GC)
Flammable
Corrosive
Irritant
N,O-Bis(trimethylsilyl)acetamide kit ≥80.0% (by GC)
Catalog # TCIAB0911-1KIT
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
Irritant
N,O-Bis(trimethylsilyl)acetamide kit ≥80.0% (by GC)
Catalog # TCIAB0911-1KIT
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    1 mL × 8

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 80.0 %
  • High boiling impurity of Silylation agent:
    No impurity higher than Isopropyl myristate (50 ppm) after 5 min + hold up time

Specification Test Results

About this item

 :
 :203.43 g/mol
Boiling Pt:73°C
Flash Pt:42°C
EINECS:233-892-7
UN:2920
ADR:8,II
 :GHS02
 :GHS05
 :GHS07
CAS Number:10416-59-8
Formula:C₈H₂₁NOSi₂
MDL Number:MFCD00008270