Bis(2-hydroxy-5-chlorophenyl)sulphide ≥98.0% (by GC, titration analysis)
TCIAB0850-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:174.0 to 177.0 deg-C
- Solubility in Methanol:almost transparency
:
:287.15 g/mol
:14,04004
:202-568-7
:GHS07
:97-24-5
:C₁₂H₈Cl₂O₂S
:MFCD00031479