Order Entry
Finland
ContactUsLinkComponent
 
 
Butyl methyl sulphide ≥98.0% (by GC)
  TCIAB0729-5ML
 :  TCI
 :  
 
Butyl methyl sulphide ≥98.0% (by GC)
  TCIAB0729-5ML
 :  TCI
 :  

 

  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :104.21 g/mol
 :123°C
 :20°C
 :211-034-2
 :1993
 :3,II
 :GHS02
 :628-29-5
 :C₅H₁₂S
 :MFCD00015256