


N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile trimethylsilylating reagent, for NH₂ compounds
Catalog # TCIAB0510-12ML
Supplier: TCI
CAS Number:
Specifications
- Pk:12 mL
- Seq:2288
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Effect test of TMS derivatization:Effective as derivatization agent DL-Threonine
Specification Test Results
About this item
:
:203.43 g/mol
Storage Temperature:Refrigerator
EINECS:233-892-7
UN:1992
ADR:3,II
:GHS02
:GHS06
:GHS08
CAS Number:10416-59-8
Formula:C₈H₂₁NOSi₂
MDL Number:MFCD00008270