Benzoyl-β-alanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3440-28-6
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD00037293
:193.20 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:131.0 to 135.0 deg-C
- Solubility in Methanol:almost transparency