Order Entry
Finland
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile (TMS-BA) for gas chromatography
Flammable
Toxic
Health
N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile (TMS-BA) for gas chromatography
Catalog # TCIAA5602-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Toxic
Health
N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile (TMS-BA) for gas chromatography
Catalog # TCIAA5602-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Effect test of TMS derivatization:
    Effective as derivatization agent DL-Threonine
  • High boiling impurity of Silylation agent:
    No impurity higher than Isopropyl myristate (50 ppm) after 5 min + hold up time

Specification Test Results

About this item

 :
 :203.43 g/mol
Storage Temperature:Refrigerator
EINECS:233-892-7
UN:1992
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:10416-59-8
Formula:C₈H₂₁NOSi₂
MDL Number:MFCD00008270