AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole) ≥95.0% (by HPLC) for HPLC labeling
Catalog # TCIAA5576-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Effect test of HPLC derivatization:Effective as labeling agent for Caproic Acid, Lauric Acid, Myristic Acid, Palmitic Acid
Specification Test Results
About this item
:
:209.22 g/mol
Storage Temperature:Refrigerator
CAS Number:254973-02-9
Formula:C₈H₇N₃O₂S
MDL Number:MFCD04038408