Order Entry
Finland
ContactUsLinkComponent
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt) ≥98.0% (by HPLC) for spectrophotometry
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt) ≥98.0% (by HPLC) for spectrophotometry
Catalog # TCIAA5107-1G
Supplier:  TCI
CAS Number:  
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt) ≥98.0% (by HPLC) for spectrophotometry
Catalog # TCIAA5107-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Dark green to Dark blue to Black powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Absorbance:
    min. 0.440 (5 mg/L, pH 5.8, 591.0 to 595.0 nm)
  • Drying loss:
    max. 7.0 %
  • Transition interval (pH):
    (Yellow green)pH3.4 to pH5.4 (Blue purple)

Specification Test Results

About this item

 :
 :700.06 g/mol
EINECS:263-661-6
CAS Number:62637-91-6
Formula:C₂₂H₁₄Br₄O₄
MDL Number:MFCD00011662