TPPS hydrate (=tetraphenylporphyrin tetrasulphonic acid hydrate) ≥85.0% (by HPLC) for spectrophotometry
Supplier: TCI
Certificates
About this item
CAS Number:35218-75-8
:
Formula:C₄₄H₃₀N₄O₁₂S₄
MDL Number:MFCD00070632
:934.98 g/mol
Specification Test Results
- Appearance:Dark blue powder to crystal
- Purity (HPLC):min. 85.0 area%
- Molar extinction coefficient:min. 430,000 (pH 7.5, 415.0 to 420.0 nm)
- Water:3.0 to 8.5 %
Specification Test Results
Frequently Bought Together