
5-Amino-1-methyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1192-21-8
:
Formula:C₄H₇N₃
MDL Number:MFCD00068156
:97.12 g/mol
Boiling Pt:105°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:gt. 69.0 and le 73.0 deg-C