Akt inhibitor VIII ≥97.0% (by HPLC)
Catalog # TCIAA3153-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):16.80 to 18.60 %
Specification Test Results
About this item
:
:551.65 g/mol
Storage Temperature:Freezer
CAS Number:612847-09-3
Formula:C₃₄H₂₉N₇O
MDL Number:MFCD09026909