5-Azoniaspiro[4.4]nonane bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16450-38-7
:
Formula:C₈H₁₆BrN
MDL Number:MFCD30749246
:206.13 g/mol
Melting Pt:273°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %