
5-Amino-2-bromo-3-methylpyridine ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:38186-83-3
:
Formula:C₆H₇BrN₂
MDL Number:MFCD03095085
:187.04 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:98.0 to 102.0 deg-C