
2-Acetamido-5-bromobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:38985-79-4
:
Formula:C₉H₈BrNO₃
MDL Number:MFCD00040904
:258.07 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:215.0 to 219.0 deg-C