
4-Acetamido-2-methylbenzoic acid ≥96.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:103204-69-9
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD02258874
:193.21 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 96.0 %