
4'-Acetamido-2'-methylacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34956-31-5
:
Formula:C₁₁H₁₃NO₂
MDL Number:MFCD02258876
:191.23 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:137.0 to 141.0 deg-C