Order Entry
Finland
ContactUsLinkComponent
 
 
3-Acetyl-2,4-dimethylpyrrole ≥98.0% (by GC)
 :  TCI
 
 
3-Acetyl-2,4-dimethylpyrrole ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    136.0 to 140.0 deg-C