
3-Amino-N-(tert-butoxycarbonyl)benzylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:147291-66-5
:
Formula:C₁₂H₁₈N₂O₂
MDL Number:MFCD04974010
:222.29 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:48.0 to 56.0 deg-C