
4-Amino-6-hydroxypyrazolo[3,4-d]pyrimidine ≥93.0% (by HPLC)
Catalog # TCIAA2699-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystalline
- Purity (HPLC):min. 93.0 area%
- Elemental analysis (Nitrogen):43.10 to 48.60 %
Specification Test Results
About this item
:
:151.13 g/mol
EINECS:226-811-1
:GHS07
CAS Number:5472-41-3
Formula:C₅H₅N₅O
MDL Number:MFCD00005689