Asiatic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:464-92-6
:
Formula:C₃₀H₄₈O₅
MDL Number:MFCD00238541
:488.709 g/mol
Melting Pt:318°C
EINECS:482-720-9
Specification Test Results
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Specific rotation [a]20/D:+47.0 to +52.0 deg (C=0.1, MeOH)