DL-ɑ-Amino-ε-caprolactam hydrochloride ≥97.0% (by total nitrogen and titration analysis)
Catalog # TCIAA2331-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
About this item
Synonyms: DL-3-Amino-2-oxohexamethyleneimine Hydrochloride, 3-Aminoazepan-2-one hydrochloride, DL-3-Aminohexahydro-2-azepinone Hydrochloride
:
:164.63 g/mol
Melting Pt:294°C
:GHS07
CAS Number:29426-64-0
Formula:C₆H₁₂N₂O
MDL Number:MFCD06798179