Order Entry
Finland
ContactUsLinkComponent
N-(Allyloxycarbonyloxy)succinimide ≥97.0% (by GC)
Irritant
N-(Allyloxycarbonyloxy)succinimide ≥97.0% (by GC)
Supplier:  TCI
Certificates
Irritant
N-(Allyloxycarbonyloxy)succinimide ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together