
4-Aminocyclohexaneethanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:857831-26-6
:
Formula:C₈H₁₇NO
MDL Number:MFCD15072144
:143.23 g/mol
Melting Pt:85°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 90.0 % (total of isomers)
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together