5-Amino-2-methoxybenzoic acid ≥97.0% (by titrimetric analysis)
Catalog # TCIAA2184-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Yellow to Amber to Dark red powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:163.0 to 167.0 deg-C
Specification Test Results
About this item
:
:167.16 g/mol
CAS Number:3403-47-2
Formula:C₈H₉NO₃
MDL Number:MFCD09260887