cis-3-Aminocyclohexanecarboxylic acid ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16636-51-4
:MFCD11042667
Formula:C₇H₁₃NO₂
MDL Number:MFCD09753647
:143.19 g/mol
Melting Pt:278°C
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
Specification Test Results
Frequently Bought Together