Order Entry
Finland
ContactUsLinkComponent
L-Alanine-tert-butyl ester hydrochloride ≥97.0% (by total nitrogen basis)
L-Alanine-tert-butyl ester hydrochloride ≥97.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
L-Alanine-tert-butyl ester hydrochloride ≥97.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Specific rotation [a]20/D:
    +1.0 to +3.0 deg (C=2, EtOH)

Specification Test Results

Related Information