
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide ≥98.0% (by titrimetric analysis)
Catalog # TCIAA1699-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:128.0 to 131.0 deg-C
- Specific rotation [a]20/D:+25.0 to +35.0 deg (C=1, CH3CN)
- Solubility in hot Acetonitrile:almost transparency
Specification Test Results
About this item
:
:366.48 g/mol
:GHS07
CAS Number:167316-27-0
Formula:C₂₁H₂₂N₂O₂S
MDL Number:MFCD03095684