Nω-(2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-β-D-glucopyranosyl)-Nɑ-(tert-butoxycarbonyl)-L-asparagine benzyl ester ≥97.0% (by HPLC)
Catalog # TCIAA1685-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):5.0 to 5.5 %
- Specific rotation [a]20/D:+47.0 to +53.0 deg (C=1,CHCl3)
Specification Test Results
About this item
:
:795.93 g/mol
Storage Temperature:Freezer
CAS Number:219968-28-2
Formula:C₄₅H₅₃N₃O₁₀
MDL Number:MFCD06797050